Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Heptyn-3-ol, 98%, Thermo Scientific™

Catalog No. AAL0533003
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0533003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0533003 Supplier Thermo Scientific Chemicals Supplier No. L0533003

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
1-Heptyn-3-ol, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used in organic synthesis.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents, heavy metal powders.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7383-19-9
Molecular Formula C7H12O
Molecular Weight (g/mol) 112.172
MDL Number MFCD00041597
InChI Key SHSFXAVQBIEYMK-UHFFFAOYSA-N
Synonym 1-heptyn-3-ol, n-butyl ethynyl carbinol, acmc-1biv8, 1-heptyn-3-ol, +/-
PubChem CID 93050
IUPAC Name hept-1-yn-3-ol
SMILES CCCCC(C#C)O

Specifications

Density 0.869
Boiling Point 161°C to 163°C
Flash Point 59°C (138°F)
Odor Recognizable
Refractive Index 1.44
Quantity 1 g
UN Number UN1987
Beilstein 1740804
Solubility Information Slightly soluble in water.
Formula Weight 112.17
Percent Purity 98%
Chemical Name or Material 1-Heptyn-3-ol
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226-H302
Flammable liquid and vapor.
Harmful if swallowed.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.