Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-heptyne, 7-bromo | 81216-14-0 | MFCD20343079 | 99.3% | 175.07 | C7H11Br | 100 MG
SDP

Catalog No. 5000281406
Encompass_Preferred

7-Bromohept-1-yne is a small-molecule PROTAC linker featuring a terminal alkyne and a primary alkyl bromide. It is used as a bifunctional building block in the synthesis of targeted protein degradation compounds and other linker-dependent organic syntheses.

  • Contains terminal alkyne and primary alkyl bromide functional groups.
  • Molecular formula C7H11Br and molecular weight 175.07.
  • CAS number 81216-14-0 for unambiguous identification.
  • High reported purity (approximately 99.3%).
  • Available in multiple pack sizes, including 100 MG and larger quantities.
  • Suitable for use as a linker in PROTAC and other synthetic chemistry workflows.
  • Provided with product datasheet and batch documentation for quality verification.

Catalog No. 50-002-81406 Supplier Medchemexpress LLC Supplier No. HYW459522100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

7-Bromohept-1-yne is a small-molecule PROTAC linker featuring a terminal alkyne and a primary alkyl bromide. It is used as a bifunctional building block in the synthesis of targeted protein degradation compounds and other linker-dependent organic syntheses.

  • Contains terminal alkyne and primary alkyl bromide functional groups.
  • Molecular formula C7H11Br and molecular weight 175.07.
  • CAS number 81216-14-0 for unambiguous identification.
  • High reported purity (approximately 99.3%).
  • Available in multiple pack sizes, including 100 MG and larger quantities.
  • Suitable for use as a linker in PROTAC and other synthetic chemistry workflows.
  • Provided with product datasheet and batch documentation for quality verification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.