Learn More
Medchemexpress LLC 1-heptyne, 7-bromo | 81216-14-0 | MFCD20343079 | 99.7% | 175.07 | C7H11Br | 500 MG
SDP
Catalog No. 5000281036
Encompass_Preferred
7-Bromohept-1-yne is a brominated terminal alkyne used as a PROTAC linker and organic building block in medicinal chemistry. Its terminal alkyne and bromo substituent enable selective coupling and functionalization steps for assembling bifunctional degraders and other small molecules.
  • Brominated terminal alkyne functionality for versatile coupling reactions.
  • Molecular formula C7H11Br and molecular weight 175.07 for synthesis calculations.
  • High reported purity: 99.7% (GC), suitable for synthetic workflows.
  • Suited for use in PROTAC linker assembly and general medicinal chemistry applications.
  • Store under recommended conditions to maintain stability.
Catalog No. 50-002-81036 Supplier Medchemexpress LLC Supplier No. HYW459522500MG
7-Bromohept-1-yne is a brominated terminal alkyne used as a PROTAC linker and organic building block in medicinal chemistry. Its terminal alkyne and bromo substituent enable selective coupling and functionalization steps for assembling bifunctional degraders and other small molecules.
  • Brominated terminal alkyne functionality for versatile coupling reactions.
  • Molecular formula C7H11Br and molecular weight 175.07 for synthesis calculations.
  • High reported purity: 99.7% (GC), suitable for synthetic workflows.
  • Suited for use in PROTAC linker assembly and general medicinal chemistry applications.
  • Store under recommended conditions to maintain stability.