Learn More
Medchemexpress LLC 1-heptyne, 7-bromo | 81216-14-0 | MFCD20343079 | 99.7% | 175.07 g/mol | C7H11Br | 1 G
SDP
Catalog No. 5000281099
Encompass_Preferred
7-Bromohept-1-yne is a bromoalkyne reagent used in medicinal chemistry as a PROTAC linker for constructing bifunctional degraders such as reported research compounds. It is provided as a research-grade material with supporting analytical documentation for laboratory synthesis and method development.
  • Bromoalkyne functional group enables chemoselective coupling reactions.
  • Designed for use as a PROTAC linker in targeted protein degradation synthesis.
  • High purity appropriate for research applications.
  • Supplied with a certificate of analysis for analytical traceability.
  • Suitable for standard laboratory storage and handling when kept under recommended conditions.
Catalog No. 50-002-81099 Supplier Medchemexpress LLC Supplier No. HYW4595221G
7-Bromohept-1-yne is a bromoalkyne reagent used in medicinal chemistry as a PROTAC linker for constructing bifunctional degraders such as reported research compounds. It is provided as a research-grade material with supporting analytical documentation for laboratory synthesis and method development.
  • Bromoalkyne functional group enables chemoselective coupling reactions.
  • Designed for use as a PROTAC linker in targeted protein degradation synthesis.
  • High purity appropriate for research applications.
  • Supplied with a certificate of analysis for analytical traceability.
  • Suitable for standard laboratory storage and handling when kept under recommended conditions.