Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Hexacosanol 95.0+%, TCI America™
SDP

Catalog No. H0342100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0342100MG 100 mg
H03421G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H0342100MG Supplier TCI America Supplier No. H0342100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 506-52-5
Molecular Formula C26H54O
Molecular Weight (g/mol) 382.717
MDL Number MFCD00002940
InChI Key IRHTZOCLLONTOC-UHFFFAOYSA-N
Synonym 1-hexacosanol, ceryl alcohol, n-hexacosanol, hexacosyl alcohol, hexacosanol, cerylic alcohol, unii-m7sd300nnb, m7sd300nnb, cerylalkohol, hexacosanyl alcohol
PubChem CID 68171
ChEBI CHEBI:28415
IUPAC Name hexacosan-1-ol
SMILES CCCCCCCCCCCCCCCCCCCCCCCCCCO

Specifications

Melting Point 80°C
Color White-Yellow
Quantity 100 mg
Formula Weight 382.72
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Hexacosanol
EINECSNumber (2)-0217
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.