Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-Hexadecanaminium, N-hexadecyl-N,N-dimethyl-, bromide (1:1) | 70755-47-4 | 99.7% | 574.86 | 25 G
SDP

Catalog No. 5000367103
Encompass_Preferred

Dicetyldimonium bromide is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is intended for research use only and not for sale to patients.

  • High purity: 99.71%
  • Molecular weight: 574.86
  • Chemical formula: C34H72BrN
  • CAS number: 70755-47-4
  • Appearance: White to off-white solid
  • Soluble in DMSO at 2 mg/mL

Catalog No. 50-003-67103 Supplier Medchemexpress LLC Supplier No. 25GHYW010731
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dicetyldimonium bromide is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is intended for research use only and not for sale to patients.

  • High purity: 99.71%
  • Molecular weight: 574.86
  • Chemical formula: C34H72BrN
  • CAS number: 70755-47-4
  • Appearance: White to off-white solid
  • Soluble in DMSO at 2 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.