Learn More
Apexbio Technology LLC 1-Hexadecanol 36653-82-4 50mg
SDP
Catalog No. 5000089812
Encompass_Preferred
1-Hexadecanol (CAS 36653-82-4) is a long-chain aliphatic fatty alcohol frequently employed as an intermediate in the synthesis of various surfactants and emollients While it does not directly target specific biological pathways its amphipathic properties enable it to modulate membrane fluidity and influence lipid organization in cellular models In biomedical research 1-hexadecanol is utilized to study membrane biophysics lipid metabolism and as a precursor in the formulation of delivery vehicles for pharmaceuticals Its role in altering membrane properties also makes it valuable for investigating cellular permeability and drug transport mechanisms
Catalog No. 50-000-89812 Supplier Apexbio Technology LLC Supplier No. B165150
1-Hexadecanol (CAS 36653-82-4) is a long-chain aliphatic fatty alcohol frequently employed as an intermediate in the synthesis of various surfactants and emollients While it does not directly target specific biological pathways its amphipathic properties enable it to modulate membrane fluidity and influence lipid organization in cellular models In biomedical research 1-hexadecanol is utilized to study membrane biophysics lipid metabolism and as a precursor in the formulation of delivery vehicles for pharmaceuticals Its role in altering membrane properties also makes it valuable for investigating cellular permeability and drug transport mechanisms