Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-Hexadecanol | 36653-82-4 | MFCD00004760 | 99.8% | 25 G
SDP

Catalog No. 5000414503
Encompass_Preferred

1-Hexadecanol is a fatty alcohol and a lipophilic substrate that can be used for ferroptosis research. It increases the level of lipid peroxidation in HT1080 cells and increases FAR1 KO HT1080 cells sensitivity to ferroptosis.

  • Increases the level of lipid peroxidation in HT1080 cells (at 20 μM, 12 h)
  • Increases FAR1 KO HT1080 cells sensitivity to ferroptosis (at 10 μM, 12 h)
  • Purity: 99.76%
  • Appearance: Solid, white to off-white
  • Initial source: Endogenous metabolite

Catalog No. 50-004-14503 Supplier Medchemexpress LLC Supplier No. 25GHYB1465
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Hexadecanol is a fatty alcohol and a lipophilic substrate that can be used for ferroptosis research. It increases the level of lipid peroxidation in HT1080 cells and increases FAR1 KO HT1080 cells sensitivity to ferroptosis.

  • Increases the level of lipid peroxidation in HT1080 cells (at 20 μM, 12 h)
  • Increases FAR1 KO HT1080 cells sensitivity to ferroptosis (at 10 μM, 12 h)
  • Purity: 99.76%
  • Appearance: Solid, white to off-white
  • Initial source: Endogenous metabolite

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.