Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-hexadecanol-d4 | 1398065-49-0 | 246.47 | C16H30D4O | 10 MG
SDP

Catalog No. 5000298040
Encompass_Preferred

1-Hexadecanol-d4 is the deuterium-labeled isotopologue of 1-hexadecanol, provided as a stable-isotope analytical standard for mass spectrometry, isotopic tracing, and related biochemical research.

  • Deuterium labeling: d4.
  • CAS: 1398065-49-0.
  • Molecular formula: C16H30D4O.
  • Molecular weight: 246.47.
  • Typical package sizes: 5 mg, 10 mg, 50 mg, 100 mg.
  • Applications: internal standard for MS quantification and metabolic tracing.

Catalog No. 50-002-98040 Supplier Medchemexpress LLC Supplier No. HYB1465S410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Hexadecanol-d4 is the deuterium-labeled isotopologue of 1-hexadecanol, provided as a stable-isotope analytical standard for mass spectrometry, isotopic tracing, and related biochemical research.

  • Deuterium labeling: d4.
  • CAS: 1398065-49-0.
  • Molecular formula: C16H30D4O.
  • Molecular weight: 246.47.
  • Typical package sizes: 5 mg, 10 mg, 50 mg, 100 mg.
  • Applications: internal standard for MS quantification and metabolic tracing.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.