Learn More
Medchemexpress LLC 1-Hexadecanol-d4 | 1398065-49-0 | 5 MG
SDP
Catalog No. 5000412676
Encompass_Preferred
1-Hexadecanol-d4 is the deuterium labeled 1-Hexadecanol, which is a fatty alcohol and a lipophilic substrate. This compound is suitable for use as a tracer or as an internal standard for quantitative analysis by methods such as NMR, GC-MS, or LC-MS.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-12676 Supplier Medchemexpress LLC Supplier No. HYB1465S45MG
1-Hexadecanol-d4 is the deuterium labeled 1-Hexadecanol, which is a fatty alcohol and a lipophilic substrate. This compound is suitable for use as a tracer or as an internal standard for quantitative analysis by methods such as NMR, GC-MS, or LC-MS.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS