Learn More
Medchemexpress LLC 1-Hexadecanol-d5 | 1219799-18-4 | 99.2% | 50 MG
SDP
Catalog No. 5000412917
Encompass_Preferred
1-Hexadecanol-d5 is a deuterium labeled version of 1-Hexadecanol, which is a fatty alcohol and a lipophilic substrate.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements are incorporated into drug molecules, primarily as tracers for quantitation during drug development.
  • Deuteration has the potential to influence the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-12917 Supplier Medchemexpress LLC Supplier No. HYB1465S250MG
1-Hexadecanol-d5 is a deuterium labeled version of 1-Hexadecanol, which is a fatty alcohol and a lipophilic substrate.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements are incorporated into drug molecules, primarily as tracers for quantitation during drug development.
  • Deuteration has the potential to influence the pharmacokinetic and metabolic profiles of drugs.