Learn More
Medchemexpress LLC 1-Hexadecanol (Palmityl alcohol) | 36653-82-4 | 99.76% | 100 G
SDP
Catalog No. 5000413379
Encompass_Preferred
1-Hexadecanol is a fatty alcohol and a lipophilic substrate, primarily utilized for research purposes. It is an endogenous metabolite and has been investigated for its role in ferroptosis research. This product is for research use only and not for sale to patients.
  • Increases the level of lipid peroxidation in HT1080 cells at a concentration of 20 μM over 12 hours.
  • Increases the sensitivity of FAR1 KO HT1080 cells to ferroptosis at a concentration of 10 μM over 12 hours.
  • Shows no effect on RSL3-induced ferroptosis in HT1080 cells expressing GNPAT or AGPS shRNA at a concentration of 20 μM over 12 hours.
Catalog No. 50-004-13379 Supplier Medchemexpress LLC Supplier No. HYB1465100G
1-Hexadecanol is a fatty alcohol and a lipophilic substrate, primarily utilized for research purposes. It is an endogenous metabolite and has been investigated for its role in ferroptosis research. This product is for research use only and not for sale to patients.
  • Increases the level of lipid peroxidation in HT1080 cells at a concentration of 20 μM over 12 hours.
  • Increases the sensitivity of FAR1 KO HT1080 cells to ferroptosis at a concentration of 10 μM over 12 hours.
  • Shows no effect on RSL3-induced ferroptosis in HT1080 cells expressing GNPAT or AGPS shRNA at a concentration of 20 μM over 12 hours.