Learn More
Medchemexpress LLC 1-Hexadecanol (Palmityl alcohol) | 36653-82-4 | 99.8% | 5 G
SDP
Catalog No. 5000412701
Encompass_Preferred
1-Hexadecanol is a fatty alcohol and a lipophilic substrate, also classified as an endogenous metabolite. It can be utilized for research related to ferroptosis, increasing lipid peroxidation in certain cells. This product is suitable for laboratory research and storage.
  • Fatty alcohol and lipophilic substrate
  • Used for ferroptosis research
  • Increases lipid peroxidation in HT1080 cells
  • Available as a white to off-white solid
  • Endogenous metabolite
Catalog No. 50-004-12701 Supplier Medchemexpress LLC Supplier No. HYB14655G
1-Hexadecanol is a fatty alcohol and a lipophilic substrate, also classified as an endogenous metabolite. It can be utilized for research related to ferroptosis, increasing lipid peroxidation in certain cells. This product is suitable for laboratory research and storage.
  • Fatty alcohol and lipophilic substrate
  • Used for ferroptosis research
  • Increases lipid peroxidation in HT1080 cells
  • Available as a white to off-white solid
  • Endogenous metabolite