Learn More
Medchemexpress LLC 1-Hexadecanol (Standard) | 36653-82-4 | 99.1% | 100 MG
SDP
Catalog No. 5000412918
Encompass_Preferred
1-Hexadecanol (Standard) is the analytical standard of 1-Hexadecanol, a fatty alcohol and a lipophilic substrate. It is intended for research and analytical applications, and can be used for the study of ferroptosis. This compound serves as a reference standard for assays and is commonly utilized in qualitative, quantitative, and methodological research experiments involving HPLC, GC, and MS.
  • Analytical standard for 1-Hexadecanol
  • Used for research and analytical applications
  • Fatty alcohol and lipophilic substrate
  • Can be used for the study of ferroptosis
  • Reference standard for assays
  • Utilized in qualitative, quantitative, and methodological research experiments involving HPLC, GC, and MS
Catalog No. 50-004-12918 Supplier Medchemexpress LLC Supplier No. HYB1465R100MG
1-Hexadecanol (Standard) is the analytical standard of 1-Hexadecanol, a fatty alcohol and a lipophilic substrate. It is intended for research and analytical applications, and can be used for the study of ferroptosis. This compound serves as a reference standard for assays and is commonly utilized in qualitative, quantitative, and methodological research experiments involving HPLC, GC, and MS.
  • Analytical standard for 1-Hexadecanol
  • Used for research and analytical applications
  • Fatty alcohol and lipophilic substrate
  • Can be used for the study of ferroptosis
  • Reference standard for assays
  • Utilized in qualitative, quantitative, and methodological research experiments involving HPLC, GC, and MS