Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (±)-1-hexadecanoyl-3-chloropropane-d5-diol | 1189718-15-7 | 98.1% | 353.98 | C19H32D5ClO3 | 10 MG
SDP

Catalog No. 5000285891
Encompass_Preferred

(±)-1-hexadecanoyl-3-chloropropane-d5-diol is a deuterium-labeled (d5), racemic reference standard of 1-hexadecanoyl-3-chloropropane-diol supplied for use as an isotopically labeled tracer and analytical standard. It is intended for applications such as quantitative mass spectrometry, pharmacokinetic studies, and metabolic tracing.

  • Deuterium-labeled (d5) racemic standard for quantitative MS and tracer studies.
  • High purity: 98.1% (assay by HPLC).
  • Molecular formula: C19H32D5ClO3; molecular weight: 353.98.
  • Appearance: white to off-white (liquid).
  • Storage: pure form -20°C (up to 3 years); in solvent -80°C (up to 6 months), -20°C (up to 1 month).

Catalog No. 50-002-85891 Supplier Medchemexpress LLC Supplier No. HY151895S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(±)-1-hexadecanoyl-3-chloropropane-d5-diol is a deuterium-labeled (d5), racemic reference standard of 1-hexadecanoyl-3-chloropropane-diol supplied for use as an isotopically labeled tracer and analytical standard. It is intended for applications such as quantitative mass spectrometry, pharmacokinetic studies, and metabolic tracing.

  • Deuterium-labeled (d5) racemic standard for quantitative MS and tracer studies.
  • High purity: 98.1% (assay by HPLC).
  • Molecular formula: C19H32D5ClO3; molecular weight: 353.98.
  • Appearance: white to off-white (liquid).
  • Storage: pure form -20°C (up to 3 years); in solvent -80°C (up to 6 months), -20°C (up to 1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.