Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Hexadecene, 92%, tech.

Catalog No. AC120500010
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC120500010 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC120500010 Supplier Thermo Scientific Chemicals Supplier No. 120500010
  • $88.00
    $70.65 / Each of 1

    Save $17.35

    20% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 629-73-2 | C16H32 | 224.43 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 629-73-2
Molecular Formula C16H32
Molecular Weight (g/mol) 224.43
MDL Number MFCD00008991
InChI Key GQEZCXVZFLOKMC-UHFFFAOYSA-N
Synonym 1-hexadecene, cetene, 1-cetene, hexadecylene-1, hexadecene, 1-n-hexadecene, cetylene, n-hexadec-1-ene, alpha-hexadecene, alpha-hexadecylene
PubChem CID 12395
ChEBI CHEBI:77507
IUPAC Name hexadec-1-ene
SMILES CCCCCCCCCCCCCCC=C

Specifications

CAS Min % 90.0
CAS Max % 100.0
Melting Point 3°C to 5°C
Color Brown-Orange to Yellow
Density 0.7830g/mL
Boiling Point 274°C
Flash Point 132°C
Infrared Spectrum Authentic
Assay Percent Range 90% min. (GC)
Packaging Glass bottle
Refractive Index 1.4390 to 1.443
Linear Formula CH3(CH2)13CH=CH2
Quantity 1 L
Beilstein 01, 226
Specific Gravity 0.783
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in alcohol,ether,petroleum and coal-tar,solvents,soluble in hydrocarbon solvents
Viscosity 3.83 mm2/s (20°C)
Formula Weight 224.43
Percent Purity 92%
Grade Technical
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material 1-Hexadecene, tech., 92%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May be fatal if swallowed and enters airways.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Do NOT induce vomiting.
EINECSNumber 211-105-8

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.