Learn More
Medchemexpress LLC 1-Hexadecyl lysophos 1mg
SDP
Catalog No. 5000204944
Encompass_Preferred
1-Hexadecyl lysophosphatidic acid is an ether analog of lysophosphatidic acid (LPA) containing a hexadecyl group in the sn-1 position LPA binds to five different G protein-coupled receptors and mediates a variety of biological responses including cell proliferation smooth muscle contraction platelet aggregation neurite contraction and cell motility
Catalog No. 50-002-04944 Supplier Medchemexpress LLC Supplier No. HYW1275021MG
1-Hexadecyl lysophosphatidic acid is an ether analog of lysophosphatidic acid (LPA) containing a hexadecyl group in the sn-1 position LPA binds to five different G protein-coupled receptors and mediates a variety of biological responses including cell proliferation smooth muscle contraction platelet aggregation neurite contraction and cell motility