Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Hexadecylamine, 90%

Catalog No. AC120510050
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC120510050 5 g Glass bottle
AC120511000 100 g Glass bottle
AC120515000 500 g Glass bottle
AC120510025 2.5 kg Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC120510050 Supplier Thermo Scientific Chemicals Supplier No. 120510050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 143-27-1 | C16H35N | 241.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143-27-1
Molecular Formula C16H35N
Molecular Weight (g/mol) 241.46
MDL Number MFCD00008158
InChI Key FJLUATLTXUNBOT-UHFFFAOYSA-N
Synonym hexadecylamine, 1-hexadecylamine, cetylamine, 1-hexadecanamine, 1-aminohexadecane, n-hexadecylamine, hexyldecylamine, palmitamine, palmitylamine, n-cetylamine
PubChem CID 8926
IUPAC Name hexadecan-1-amine
SMILES CCCCCCCCCCCCCCCCN

Specifications

Melting Point 38.0°C to 47.0°C
Color White
Boiling Point 330.0°C
Flash Point 140°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)15NH2
Quantity 5 g
Beilstein 04,202
Solubility Information Solubility in water: insoluble
Formula Weight 241.46
Percent Purity 90%
Physical Form Low Melting Crystalline Mass or Chunks
Chemical Name or Material 1-Hexadecylamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
EINECSNumber 205-596-8

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.