Learn More
Medchemexpress LLC 1-hexanol | 111-27-3 | MFCD00002982 | 99.4% | 102.18 | C6H14O | 25 G
SDP
Catalog No. 5000303023
Encompass_Preferred
1-Hexanol is a primary straight-chain alcohol used as a solvent and reagent in organic synthesis, extractions, and formulation work. Provided at high purity (~99.35%), it is a colorless liquid with moderate volatility and is suitable for laboratory-scale reactions and analytical applications.
  • High purity (~99.35%) suitable for analytical and synthetic work.
  • Acts as an effective solvent for organic reactions and extractions.
  • Compatible with common organic solvents and reagents.
  • Available in small pack sizes convenient for laboratory use.
  • Documentation available: data sheet, COA, and SDS for safe handling.
Catalog No. 50-003-03023 Supplier Medchemexpress LLC Supplier No. 25GHYW032022
1-Hexanol is a primary straight-chain alcohol used as a solvent and reagent in organic synthesis, extractions, and formulation work. Provided at high purity (~99.35%), it is a colorless liquid with moderate volatility and is suitable for laboratory-scale reactions and analytical applications.
  • High purity (~99.35%) suitable for analytical and synthetic work.
  • Acts as an effective solvent for organic reactions and extractions.
  • Compatible with common organic solvents and reagents.
  • Available in small pack sizes convenient for laboratory use.
  • Documentation available: data sheet, COA, and SDS for safe handling.