Learn More
Medchemexpress LLC 1-hexanol | 111-27-3 | MFCD00002982 | 99.4% | 102.18 g/mol | C6H14O | 1 KG
SDP
Catalog No. 5000301555
Encompass_Preferred
1-Hexanol (hexan-1-ol) is a primary aliphatic alcohol commonly used as a solvent, reagent, and analytical standard in chemical and biological research. Supplied as an analytical-grade material, it has purity 99.35%, molecular formula C6H14O, and molecular weight 102.18 g/mol. Available in multiple pack sizes up to 1 kg.
  • Primary alcohol with solvent and surfactant properties.
  • High purity (99.35%) suitable for analytical and chromatographic use.
  • Consistent molecular composition (C6H14O, 102.18 g/mol) for reference standards.
  • Applicable as a reagent or intermediate in organic synthesis.
  • Available in multiple pack sizes, including 1 kg, for laboratory and bulk use.
Catalog No. 50-003-01555 Supplier Medchemexpress LLC Supplier No. HYW0320221KG
1-Hexanol (hexan-1-ol) is a primary aliphatic alcohol commonly used as a solvent, reagent, and analytical standard in chemical and biological research. Supplied as an analytical-grade material, it has purity 99.35%, molecular formula C6H14O, and molecular weight 102.18 g/mol. Available in multiple pack sizes up to 1 kg.
  • Primary alcohol with solvent and surfactant properties.
  • High purity (99.35%) suitable for analytical and chromatographic use.
  • Consistent molecular composition (C6H14O, 102.18 g/mol) for reference standards.
  • Applicable as a reagent or intermediate in organic synthesis.
  • Available in multiple pack sizes, including 1 kg, for laboratory and bulk use.