Learn More
Medchemexpress LLC 1-hexanol | 111-27-3 | MFCD00002982 | 99.4% | 102.18 g/mol | C6H14O | 1 ML
SDP
Catalog No. 5000302075
Encompass_Preferred
1-Hexanol (CAS 111-27-3) is a primary aliphatic alcohol used as a laboratory reagent, solvent, and surfactant. It is supplied both as a high-purity bulk liquid and as a ready-to-use 10 mM solution in ethanol, making it suitable for biochemical studies and general research applications that require a consistent, characterized reagent.
  • High purity for research applications.
  • Available as 10 mM solution in ethanol and in bulk liquid sizes (25 g-1 kg).
  • Acts as a solvent and surfactant in experimental protocols.
  • Stable under standard laboratory storage conditions.
  • Well-characterized identity (CAS 111-27-3) for traceability.
Catalog No. 50-003-02075 Supplier Medchemexpress LLC Supplier No. HYW03202210MM/1ML
1-Hexanol (CAS 111-27-3) is a primary aliphatic alcohol used as a laboratory reagent, solvent, and surfactant. It is supplied both as a high-purity bulk liquid and as a ready-to-use 10 mM solution in ethanol, making it suitable for biochemical studies and general research applications that require a consistent, characterized reagent.
  • High purity for research applications.
  • Available as 10 mM solution in ethanol and in bulk liquid sizes (25 g-1 kg).
  • Acts as a solvent and surfactant in experimental protocols.
  • Stable under standard laboratory storage conditions.
  • Well-characterized identity (CAS 111-27-3) for traceability.