Learn More
Medchemexpress LLC 1-hexanol | 111-27-3 | MFCD00002982 | 99.4% | 102.18 g/mol | C6H14O | 100 G
SDP
Catalog No. 5000300084
Encompass_Preferred
1-Hexanol is a straight-chain primary alcohol (C6H14O) used as a solvent, surfactant, and reagent in organic synthesis and analytical workflows. It is provided as a high-purity reagent suitable for laboratory research, formulation studies, and analytical standards.
  • High purity (≈99.35%) for reliable analytical and synthetic results.
  • Effective solvent for organic reactions and liquid-liquid extractions.
  • Useful surfactant for surface and interfacial property studies.
  • Available in multiple pack sizes to support small-scale and bulk use.
  • Clear, colorless liquid with well-documented physical properties.
Catalog No. 50-003-00084 Supplier Medchemexpress LLC Supplier No. HYW032022100G
1-Hexanol is a straight-chain primary alcohol (C6H14O) used as a solvent, surfactant, and reagent in organic synthesis and analytical workflows. It is provided as a high-purity reagent suitable for laboratory research, formulation studies, and analytical standards.
  • High purity (≈99.35%) for reliable analytical and synthetic results.
  • Effective solvent for organic reactions and liquid-liquid extractions.
  • Useful surfactant for surface and interfacial property studies.
  • Available in multiple pack sizes to support small-scale and bulk use.
  • Clear, colorless liquid with well-documented physical properties.