Learn More
Medchemexpress LLC HY-W032022 500mg Medchemexpress, 1-Hexanol CAS:111-27-3 Purity:>98%
Supplier: Medchemexpress LLC HYW032022500MG
Medchemexpress, HY-W032022 500mg 1-Hexanol CAS:111-27-3 1-Hexanol, a primary alcohol, is a surfactant that can be employed in industrial processes to enhance interfacial properties. 1-Hexanol uncouples mitochondrial respiration by a non-protonophoric mechanism. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.