Learn More
Medchemexpress LLC 1-hexanol, 6-bromo- | 4286-55-9 | MFCD00002983 | 96.7% | 181.07 g/mol | C6H13BrO | 50 G
SDP
Catalog No. 5000303367
Encompass_Preferred
6-Bromo-1-hexanol is a brominated primary alcohol used as an intermediate and reagent in organic synthesis. It contains a terminal bromide that serves as a good leaving group and a primary hydroxyl that enables further derivatization; common applications include nucleophilic substitution, alkylation, and cross-linking in medicinal and polymer chemistry.
  • Terminal bromide suitable for nucleophilic substitution reactions.
  • Primary hydroxyl enables further functionalization and derivatization.
  • Serves as a versatile building block for medicinal and polymer synthesis.
  • Offered in multiple pack sizes to support different scales of work.
  • Quality-controlled with datasheet and certificate of analysis available.
Catalog No. 50-003-03367 Supplier Medchemexpress LLC Supplier No. HYW01645550G
6-Bromo-1-hexanol is a brominated primary alcohol used as an intermediate and reagent in organic synthesis. It contains a terminal bromide that serves as a good leaving group and a primary hydroxyl that enables further derivatization; common applications include nucleophilic substitution, alkylation, and cross-linking in medicinal and polymer chemistry.
  • Terminal bromide suitable for nucleophilic substitution reactions.
  • Primary hydroxyl enables further functionalization and derivatization.
  • Serves as a versatile building block for medicinal and polymer synthesis.
  • Offered in multiple pack sizes to support different scales of work.
  • Quality-controlled with datasheet and certificate of analysis available.