Learn More
Medchemexpress LLC 1-Hexanol, 6-chloro- | 2009-83-8 | 99.5% | 136.62 | 100 G
SDP
Catalog No. 5000404564
Encompass_Preferred
6-Chloro-1-hexanol is a biochemical reagent that serves as a biological material or organic compound for life science research. It is useful in the preparation of its corresponding bromide by reaction with sodium bromide, and also for the selective elimination of 6-chloro-1-hexene from esters of 6-chloro-1-hexanol.
  • Biochemical reagent
  • Used as a biological material or organic compound for life science research
  • Used in the preparation of its corresponding bromide
  • Utilized in the selective elimination of 6-chloro-1-hexene from esters
Catalog No. 50-004-04564 Supplier Medchemexpress LLC Supplier No. HY41681100G
6-Chloro-1-hexanol is a biochemical reagent that serves as a biological material or organic compound for life science research. It is useful in the preparation of its corresponding bromide by reaction with sodium bromide, and also for the selective elimination of 6-chloro-1-hexene from esters of 6-chloro-1-hexanol.
  • Biochemical reagent
  • Used as a biological material or organic compound for life science research
  • Used in the preparation of its corresponding bromide
  • Utilized in the selective elimination of 6-chloro-1-hexene from esters