Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Hexanol, 98%, Reagent Grade, Solstice

Catalog No. 60048031 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-048-031 2.5 L Glass bottle
60-048-030 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-048-031 Supplier Solstice Supplier No. H133032.5L
Add to Cart
Add to Cart

Reagent Grade, 98%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-27-3
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.177
MDL Number MFCD00002982
InChI Key ZSIAUFGUXNUGDI-UHFFFAOYSA-N
Synonym 1-hexanol, hexyl alcohol, hexanol, n-hexanol, n-hexyl alcohol, 1-hexyl alcohol, 1-hydroxyhexane, amylcarbinol, caproyl alcohol, pentylcarbinol
PubChem CID 8103
ChEBI CHEBI:87393
IUPAC Name hexan-1-ol
SMILES CCCCCCO

Specifications

Melting Point -52°C (literature)
Color Colorless
Density 0.814g/mL at 25°C
Boiling Point 156°C to 157°C (literature)
Packaging Glass bottle
Linear Formula CH3(CH2)5OH
Quantity 2.5 L
UN Number UN2282
Beilstein 969167
Vapor Pressure 1mmHg (25.6°C)
Formula Weight 102.17g/mol
Percent Purity 0.98
Chemical Name or Material 1-Hexanol
Show More Show Less
Hazard Statement P280-P305 + P351 + P338
DOTInformation 3/PGIII
RTECSNumber 203-852-3
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.