Learn More
Medchemexpress LLC 1-Hexanol-d11 | 2159-18-4 | 99.3% | 113.24 | C6H3D11O | 50 MG
SDP
Catalog No. 5000299408
Encompass_Preferred
1-Hexanol-d11 is the deuterium-labeled analogue of 1-hexanol used as an isotopic standard and tracer in analytical and research applications. Deuterium substitution produces a predictable mass shift for mass spectrometry and metabolic labeling studies; the compound is supplied as a colorless to light yellow liquid with high reported purity.
  • Deuterium-labeled isotopologue for mass-shifted analysis.
  • High purity suitable for analytical standards.
  • Useful as an internal standard in mass spectrometry workflows.
  • Stable when stored under recommended low-temperature conditions.
  • Available in small research quantities for analytical applications.
Catalog No. 50-002-99408 Supplier Medchemexpress LLC Supplier No. HYW032022S150MG
1-Hexanol-d11 is the deuterium-labeled analogue of 1-hexanol used as an isotopic standard and tracer in analytical and research applications. Deuterium substitution produces a predictable mass shift for mass spectrometry and metabolic labeling studies; the compound is supplied as a colorless to light yellow liquid with high reported purity.
  • Deuterium-labeled isotopologue for mass-shifted analysis.
  • High purity suitable for analytical standards.
  • Useful as an internal standard in mass spectrometry workflows.
  • Stable when stored under recommended low-temperature conditions.
  • Available in small research quantities for analytical applications.