Learn More
Medchemexpress LLC 1-hexanol-d11 | 2159-18-4 | MFCD08705002 | 99.3% | 113.24 | C6H3D11O | 10 MG
SDP
Catalog No. 5000301769
Encompass_Preferred
1-Hexanol-d11 is the deuterium-labeled analogue of 1-hexanol supplied for analytical and research applications. It is intended for use as an isotope-labeled reagent or internal standard in mass spectrometry, NMR, and tracer studies, providing spectral clarity and reliable quantitation.
  • High isotopic purity supporting accurate mass spectrometry and tracer experiments.
  • High chemical purity to minimize interfering signals in analyses.
  • Suitable as an internal standard for quantitative workflows.
  • Compatible with MS and NMR analytical techniques.
  • Available in multiple small pack sizes for method development and routine use.
  • Specified storage conditions to preserve long-term stability.
Catalog No. 50-003-01769 Supplier Medchemexpress LLC Supplier No. HYW032022S110MG
1-Hexanol-d11 is the deuterium-labeled analogue of 1-hexanol supplied for analytical and research applications. It is intended for use as an isotope-labeled reagent or internal standard in mass spectrometry, NMR, and tracer studies, providing spectral clarity and reliable quantitation.
  • High isotopic purity supporting accurate mass spectrometry and tracer experiments.
  • High chemical purity to minimize interfering signals in analyses.
  • Suitable as an internal standard for quantitative workflows.
  • Compatible with MS and NMR analytical techniques.
  • Available in multiple small pack sizes for method development and routine use.
  • Specified storage conditions to preserve long-term stability.