Learn More
Medchemexpress LLC 1-hexanol-d13 | 204244-84-8 | 98.8% | 115.25 g·mol-1 | C6HD13O | 10 MG
SDP
Catalog No. 5000299856
Encompass_Preferred
1-Hexanol-d13 is the deuterium-labeled analogue of 1-hexanol provided as a high-purity liquid reference standard for analytical and research applications. It is intended for use in mass spectrometry, isotope tracing, and metabolic studies where deuterium labeling enables precise internal standardization and tracer experiments.
  • High purity (98.8%) suitable for analytical applications.
  • Deuterium-labeled for isotope tracing and internal standard use.
  • Supplied as a liquid, colorless to light yellow.
  • Available in small pack sizes for research-scale experiments.
  • Delivered with datasheet, SDS, and COA for traceability.
Catalog No. 50-002-99856 Supplier Medchemexpress LLC Supplier No. HYW032022S10MG
1-Hexanol-d13 is the deuterium-labeled analogue of 1-hexanol provided as a high-purity liquid reference standard for analytical and research applications. It is intended for use in mass spectrometry, isotope tracing, and metabolic studies where deuterium labeling enables precise internal standardization and tracer experiments.
  • High purity (98.8%) suitable for analytical applications.
  • Deuterium-labeled for isotope tracing and internal standard use.
  • Supplied as a liquid, colorless to light yellow.
  • Available in small pack sizes for research-scale experiments.
  • Delivered with datasheet, SDS, and COA for traceability.