Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-Hexanol-d13 | 204244-84-8 | MFCD00144898 | 98.8% | 115.25 g/mol | C6HD13O | 5 MG
SDP

Catalog No. 5000299591
Encompass_Preferred

1-Hexanol-d13 is the deuterium-labeled analogue of 1-hexanol used primarily as an internal standard and tracer for quantitative analytical methods. It enables accurate measurement in NMR, GC-MS, and LC-MS workflows and supports metabolite and pharmacokinetic studies.

  • Deuterium-labeled tracer for accurate quantitation.
  • Purity 98.8% for reliable analytical performance.
  • Molecular weight 115.25 g/mol and formula C6HD13O.
  • Liquid, colorless to light yellow physical state.
  • Stable under recommended storage conditions for long-term use.
  • Suitable for NMR, GC-MS, and LC-MS internal standard applications.
  • Useful for metabolite profiling and pharmacokinetic studies.

Catalog No. 50-002-99591 Supplier Medchemexpress LLC Supplier No. HYW032022S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Hexanol-d13 is the deuterium-labeled analogue of 1-hexanol used primarily as an internal standard and tracer for quantitative analytical methods. It enables accurate measurement in NMR, GC-MS, and LC-MS workflows and supports metabolite and pharmacokinetic studies.

  • Deuterium-labeled tracer for accurate quantitation.
  • Purity 98.8% for reliable analytical performance.
  • Molecular weight 115.25 g/mol and formula C6HD13O.
  • Liquid, colorless to light yellow physical state.
  • Stable under recommended storage conditions for long-term use.
  • Suitable for NMR, GC-MS, and LC-MS internal standard applications.
  • Useful for metabolite profiling and pharmacokinetic studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.