Learn More
Medchemexpress LLC 1-hexanol-d3 | 52598-04-6 | >98.0% | 104.19 g/mol | C6H12D2O | 1 MG
SDP
Catalog No. 5000299569
Encompass_Preferred
1-Hexanol-d3 is the deuterium-labeled isotopologue of 1-hexanol used as a stable isotope standard in analytical workflows. It retains the chemical behavior of 1-hexanol while providing a mass difference for reliable mass spectrometry quantitation (CAS 52598-04-6).
  • Deuterium-labeled internal standard for GC-MS and LC-MS.
  • Suitable for quantitative analysis and method validation.
  • Chemical behavior matches native 1-hexanol.
  • High isotopic purity (>98% D) for accurate isotope-based measurements.
  • Supplied in small analytical quantities suitable for standards preparation.
Catalog No. 50-002-99569 Supplier Medchemexpress LLC Supplier No. HYW032022S21MG
1-Hexanol-d3 is the deuterium-labeled isotopologue of 1-hexanol used as a stable isotope standard in analytical workflows. It retains the chemical behavior of 1-hexanol while providing a mass difference for reliable mass spectrometry quantitation (CAS 52598-04-6).
  • Deuterium-labeled internal standard for GC-MS and LC-MS.
  • Suitable for quantitative analysis and method validation.
  • Chemical behavior matches native 1-hexanol.
  • High isotopic purity (>98% D) for accurate isotope-based measurements.
  • Supplied in small analytical quantities suitable for standards preparation.