Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-hexanol-d3 | 52598-04-6 | 99.1% | 104.19 | C6H12D2O | 5 MG
SDP

Catalog No. 5000299407
Encompass_Preferred

1-Hexanol-d3 (CAS 52598-04-6) is the deuterium-labeled isotopologue of 1-hexanol, supplied for use as a stable-isotope reference standard in analytical chemistry and mass spectrometry. It is intended for use as an internal standard, tracer, or reference in quantitative and qualitative analyses.

  • Deuterium-labeled (d3) isotopologue for isotope-dilution and MS applications.
  • High purity: 99.1% (as listed by supplier).
  • Chemical formula C6H12D2O; molecular weight 104.19.
  • Available in small pack sizes suitable for analytical standards.
  • Storage: pure form -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-99407 Supplier Medchemexpress LLC Supplier No. HYW032022S25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Hexanol-d3 (CAS 52598-04-6) is the deuterium-labeled isotopologue of 1-hexanol, supplied for use as a stable-isotope reference standard in analytical chemistry and mass spectrometry. It is intended for use as an internal standard, tracer, or reference in quantitative and qualitative analyses.

  • Deuterium-labeled (d3) isotopologue for isotope-dilution and MS applications.
  • High purity: 99.1% (as listed by supplier).
  • Chemical formula C6H12D2O; molecular weight 104.19.
  • Available in small pack sizes suitable for analytical standards.
  • Storage: pure form -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.