Learn More
Medchemexpress LLC 1-hexanol-d3 | 52598-04-6 | 99.1% | 104.19 g/mol | C6H12D2O | 10 MG
SDP
Catalog No. 5000301772
Encompass_Preferred
1-Hexanol-d3 is a deuterium-labeled form of 1-hexanol supplied as a stable-isotope reference standard for analytical laboratories. It is intended for use as an internal standard, tracer, or reference material in mass spectrometry and other analytical techniques requiring isotopically labeled compounds.
  • Deuterium-labeled reference standard suitable for analytical applications.
  • High listed purity for reliable quantitation and trace analysis.
  • Available in small pack sizes for low-consumption analytical workflows.
  • Compatible with mass spectrometry and chromatographic methods as an internal standard.
  • Provides defined molecular weight and formula for accurate mass assignments.
Catalog No. 50-003-01772 Supplier Medchemexpress LLC Supplier No. HYW032022S210MG
1-Hexanol-d3 is a deuterium-labeled form of 1-hexanol supplied as a stable-isotope reference standard for analytical laboratories. It is intended for use as an internal standard, tracer, or reference material in mass spectrometry and other analytical techniques requiring isotopically labeled compounds.
  • Deuterium-labeled reference standard suitable for analytical applications.
  • High listed purity for reliable quantitation and trace analysis.
  • Available in small pack sizes for low-consumption analytical workflows.
  • Compatible with mass spectrometry and chromatographic methods as an internal standard.
  • Provides defined molecular weight and formula for accurate mass assignments.