Learn More
Medchemexpress LLC 1-hexanol-d5 | 64118-18-9 | 107.21 g/mol | C6H9D5O | 1 MG
SDP
Catalog No. 5000299374
Encompass_Preferred
1-Hexanol-d5 is the deuterium-labeled analogue of 1-hexanol used as a tracer and internal standard for quantitative analysis by NMR, GC-MS, and LC-MS. It is intended for analytical and research use; consult the certificate of analysis for lot-specific purity and handling instructions.
  • Deuterium-labeled internal standard for quantitative analysis.
  • Suitable for NMR, GC-MS, and LC-MS applications.
  • Provided in small pack sizes for analytical use.
  • Molecular formula C6H9D5O and molecular weight 107.21 g/mol.
  • Use as a tracer or internal standard in metabolic and analytical studies.
Catalog No. 50-002-99374 Supplier Medchemexpress LLC Supplier No. HYW032022S31MG
1-Hexanol-d5 is the deuterium-labeled analogue of 1-hexanol used as a tracer and internal standard for quantitative analysis by NMR, GC-MS, and LC-MS. It is intended for analytical and research use; consult the certificate of analysis for lot-specific purity and handling instructions.
  • Deuterium-labeled internal standard for quantitative analysis.
  • Suitable for NMR, GC-MS, and LC-MS applications.
  • Provided in small pack sizes for analytical use.
  • Molecular formula C6H9D5O and molecular weight 107.21 g/mol.
  • Use as a tracer or internal standard in metabolic and analytical studies.