Learn More
Medchemexpress LLC 1-hexanol-d5 | 64118-18-9 | >98.0% | 107.21 g/mol | C6H9D5O | 5 MG
SDP
Catalog No. 5000299375
Encompass_Preferred
1-Hexanol-d5 is the deuterium-labeled analogue of 1-hexanol supplied as a liquid stable isotope reagent for analytical and tracer applications. It is commonly used as an internal standard in mass spectrometry, quantitative analyses, and metabolic tracing where deuterium labeling improves selectivity and detection.
  • Deuterium-labeled internal standard for mass spectrometry.
  • High isotopic enrichment for reliable tracer and quantitation.
  • Liquid physical form suitable for solution preparation.
  • Stable under recommended storage conditions for long-term use.
  • Available in small, pre-weighed pack sizes for analytical workflows.
Catalog No. 50-002-99375 Supplier Medchemexpress LLC Supplier No. HYW032022S35MG
1-Hexanol-d5 is the deuterium-labeled analogue of 1-hexanol supplied as a liquid stable isotope reagent for analytical and tracer applications. It is commonly used as an internal standard in mass spectrometry, quantitative analyses, and metabolic tracing where deuterium labeling improves selectivity and detection.
  • Deuterium-labeled internal standard for mass spectrometry.
  • High isotopic enrichment for reliable tracer and quantitation.
  • Liquid physical form suitable for solution preparation.
  • Stable under recommended storage conditions for long-term use.
  • Available in small, pre-weighed pack sizes for analytical workflows.