Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Hexen-3-ol 97.0+%, TCI America™
SDP

Catalog No. H065925ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H065925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H065925ML Supplier TCI America Supplier No. H065925ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4798-44-1
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.161
MDL Number MFCD00004581
InChI Key BVOSSZSHBZQJOI-UHFFFAOYSA-N
Synonym 1-hexen-3-ol, propylvinylcarbinol, 3-hydroxy-1-hexene, propyl vinyl carbinol, vinyl propyl carbinol, 1-vinylbutanol, 1-vinylbutan-1-ol, fema no. 3608, 1-hexene-3-ol, 1-hexen-3-ol, r
PubChem CID 20928
IUPAC Name hex-1-en-3-ol
SMILES CCCC(C=C)O

Specifications

Color Yellow
Boiling Point 134°C
Quantity 25 mL
UN Number 1987
Formula Weight 100.16
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Hexen-3-ol
EINECSNumber (2)-2393
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.