Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Hexene

Catalog No. 2305451L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-054-51L 1 L
23-054-5100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-054-51L Supplier Sigma Aldrich Supplier No. 2305451L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1-Hexene is primarily used in the synthesis of poly(1-hexene) and a variety of polyethylene copolymers.

Specifications

CAS 592-41-6
Density 0.678 g/mL (at 25°C (literature))
Boiling Point 60°C to 66°C (lit.)
Molecular Formula C6H12
Refractive Index n20/D 1.388 (literature)
Linear Formula CH3(CH2)3 CH=CH2
MDL Number MFCD00009505
Quantity 1 L
Molecular Weight (g/mol) 84.16
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.