Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Hexene

Catalog No. 24076150ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
50 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-076-150ML 50 mL
24-076-1100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 24-076-150ML Supplier Sigma Aldrich Supplier No. 24076150ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • The kinetics of polymerization of 1-hexene was studied using a family of five zirconium amine bis-phenolate catalysts
  • Polymerization reaction of 1-hexene catalyzed by a series of hafnium complexes was studied

Specifications

CAS 592-41-6
Density 0.678 g/mL (at 25°C (literature))
Boiling Point 60°C to 66°C (lit.)
Molecular Formula C6H12
Refractive Index n20/D 1.388 (literature)
Linear Formula CH3(CH2)3 CH=CH2
MDL Number MFCD00009505
Quantity 50 mL
Molecular Weight (g/mol) 84.16
Percent Purity ≥99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.