Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Hexyl-2,3-dimethylimidazolium Iodide 98.0+%, TCI America™
SDP

Catalog No. H12865G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H12865G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H12865G Supplier TCI America Supplier No. H12865G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 288627-94-1
Molecular Formula C11H21IN2
Molecular Weight (g/mol) 308.207
InChI Key YYXZQUOJBJOARI-UHFFFAOYSA-M
PubChem CID 53384397
IUPAC Name 1-hexyl-2,3-dimethylimidazol-3-ium;iodide
SMILES CCCCCCN1C=C[N+](=C1C)C.[I-]

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 308.21
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Hexyl-2,3-dimethylimidazolium Iodide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.