Learn More
Accela Chembio Inc 1-hexyl-3-methylimidazolium Tetrafluoroborate | 5g | 244193-50-8 | MFCD03095459 | 97+% | D: 1.149 | Shelf Life: 1440 Days | Moisture Sensitive/n2
Catalog No. 50252067
Encompass_Preferred
1-hexyl-3-methylimidazolium Tetrafluoroborate | 5g | 244193-50-8 | MFCD03095459 | 97+% | D: 1.149 | Shelf Life: 1440 Days | Moisture Sensitive/n2
Catalog No. 50-252-067 Supplier Accela Chembio Inc Supplier No. SY0150545G
1-hexyl-3-methylimidazolium Tetrafluoroborate | 5g | 244193-50-8 | MFCD03095459 | 97+% | D: 1.149 | Shelf Life: 1440 Days | Moisture Sensitive/n2