Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Hexyl-3-Methylimidazolium Bis(Trifluormethylsulfonyl)Imide

Catalog No. 7279545G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
72-795-45G 5 g
72-795-450G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 72-795-45G Supplier Sigma Aldrich Supplier No. 7279545G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1-Hexyl-3-methylimidazolium bis(trifluormethylsulfonyl)imide is a 1-alkyl-3-methylimidazolium based ionic liquid. The solubility of gases such as oxygen and methane in HMIM TFS can be improved in the presence of carbon dioxide.

Specifications

CAS 382150-50-7
Molecular Formula C12H19F6N3O4S2
Linear Formula C12H19F6N3O4S2
MDL Number MFCD08458931
Quantity 5 g
Synonym HMIM BTI; HMIM TFS
Molecular Weight (g/mol) 447.42 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.