Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Hexyl-3-Methylimidazolium Chloride

Catalog No. 8792950G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 g
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
87-929-50G 50 mL
87-929-5G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 87-929-50G Supplier Sigma Aldrich Supplier No. 8792950G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1-hexyl-3-methylimidazolium chloride is an ionic liquid (IL). Its surface and bulk properties in aqueous solution at various temperatures indicates that it behaves as a short-chain cationic surfactant and shows aggregation behavior.Ionic liquid employed in the catalytic, solvent-free alkylation; catalyst for polymerization of olefins.

Specifications

CAS 17756-58-0
Molecular Formula C32H69NO
Linear Formula [CH3(CH2)7]4N(OH)
MDL Number MFCD00059257
Quantity 50 mL
Molecular Weight (g/mol) 483.9 g/mol
Biological Activity Eyes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.