Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Hexyl-3-Methylimidazolium Hexafluorophosphate

Catalog No. 893205G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-320-5G 50 g
89-320-50G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 89-320-5G Supplier Sigma Aldrich Supplier No. 893205G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

https://www.sigmaaldrich.com/content/dam/sigma-aldrich/materials-science/material-matters/material_matters_v4n4.pdf#page=18HMIMPF6 can be used as a chelate extraction solvent for divalent metal cations using 4,4,4-trifluoro-1-(2-thienyl)-1,3-butanedione (thenoyltrifluoroacetone, Htta) as an extractant.Ionic liquid employed in Friedel-Crafts alkylation of aromatic compounds; polarity of salt.

Specifications

CAS 304680-35-1
Molecular Formula C10H19F6N2P
Refractive Index n20/D 1.419
Linear Formula C10H19F6N2P
MDL Number MFCD03093296
Quantity 50 g
Synonym HMIMPF6
Molecular Weight (g/mol) 312.24 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.