Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Hexyl-3-methylimidazolium Iodide 98.0+%, TCI America™
SDP

Catalog No. H16335G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H16335G 5 g
H163325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H16335G Supplier TCI America Supplier No. H16335G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 178631-05-5
Molecular Formula C10H19IN2
Molecular Weight (g/mol) 294.18
MDL Number MFCD12964976
InChI Key CZIUVCSYOGFUPH-UHFFFAOYSA-M
Synonym 1-hexyl-3-methylimidazolium iodide, 1-hexyl-3-methyl-1h-imidazol-3-ium iodide, 1-n-hexyl-3-methylimidazolium iodide, dsstox_cid_27927, dsstox_rid_82680, dsstox_gsid_47951, 1-methyl-3-hexylimidazolium iodide, 1-hexyl-3-me-thylimidazolium iodide, 1h-imidazolium,1-hexyl-3-methyl-,iodide, 1-hexyl-3-methylimidazolium iodide hplc
PubChem CID 12036651
IUPAC Name 1-hexyl-3-methylimidazol-3-ium;iodide
SMILES CCCCCCN1C=C[N+](=C1)C.[I-]

Specifications

Color Brown-Yellow
Quantity 5 g
Formula Weight 294.18
Percent Purity ≥98.0% (HPLC,N)
Physical Form Liquid
Chemical Name or Material 1-Hexyl-3-methylimidazolium Iodide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.