Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Hydrazinophthalazine Hydrochloride 99.0+%, TCI America™
SDP

Catalog No. H04095G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H04095G 5 g
H040925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H04095G Supplier TCI America Supplier No. H04095G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 304-20-1
Molecular Formula C8H9ClN4
Molecular Weight (g/mol) 196.638
MDL Number MFCD00135998
InChI Key ZUXNZUWOTSUBMN-UHFFFAOYSA-N
Synonym hydralazine hydrochloride, 1-hydrazinophthalazine hydrochloride, 1-hydrazinylphthalazine hydrochloride, apresoline, aiselazine, hydralazine hcl, hydralazine chloride, hyperazin, hyperex, lopress
PubChem CID 9351
ChEBI CHEBI:31672
IUPAC Name phthalazin-1-ylhydrazine;hydrochloride
SMILES C1=CC=C2C(=C1)C=NN=C2NN.Cl

Specifications

Melting Point 273°C
Color White-Yellow
Quantity 5 g
UN Number 2811
Formula Weight 196.64
Percent Purity ≥99.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Hydrazinophthalazine Hydrochloride
RTECSNumber TH9000000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.