Learn More
Medchemexpress LLC 1-Hydroxy-2-butanone | 5077-67-8 | 88.11 | 1 G
SDP
Catalog No. 5000445988
Encompass_Preferred
1-Hydroxy-2-butanone is a natural compound isolated from Bamboo Juice. It serves as a key intermediate for ethambutol, a compound known for its anti-tuberculosis effects. This product is intended for research use only.
  • Natural compound isolated from Bamboo Juice
  • Key intermediate for ethambutol
  • Ethambutol has anti-tuberculosis effects
  • For research use only
Catalog No. 50-004-45988 Supplier Medchemexpress LLC Supplier No. HYW0053271G
1-Hydroxy-2-butanone is a natural compound isolated from Bamboo Juice. It serves as a key intermediate for ethambutol, a compound known for its anti-tuberculosis effects. This product is intended for research use only.
  • Natural compound isolated from Bamboo Juice
  • Key intermediate for ethambutol
  • Ethambutol has anti-tuberculosis effects
  • For research use only