Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-Hydroxy-2-butanone | 5077-67-8 | 88.11 | 10 G
SDP

Catalog No. 5000445497
Encompass_Preferred

1-Hydroxy-2-butanone is a natural compound isolated from Bamboo Juice. It acts as a key intermediate for ethambutol, which has anti-tuberculosis effects.

  • Purity: 97.38%
  • Molecular weight: 88.11
  • Molecular formula: C₄H₈O₂
  • CAS number: 5077-67-8
  • Appearance: Colorless to light yellow liquid (density: 1.0272 g/cm³)
  • Structure classification: Ketones, aldehydes, acids
  • Initial source: Microorganisms
  • Endogenous metabolite
  • Related to natural products, disease research, infection, and bacterial infection

Catalog No. 50-004-45497 Supplier Medchemexpress LLC Supplier No. HYW00532710G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Hydroxy-2-butanone is a natural compound isolated from Bamboo Juice. It acts as a key intermediate for ethambutol, which has anti-tuberculosis effects.

  • Purity: 97.38%
  • Molecular weight: 88.11
  • Molecular formula: C₄H₈O₂
  • CAS number: 5077-67-8
  • Appearance: Colorless to light yellow liquid (density: 1.0272 g/cm³)
  • Structure classification: Ketones, aldehydes, acids
  • Initial source: Microorganisms
  • Endogenous metabolite
  • Related to natural products, disease research, infection, and bacterial infection

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.