Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-Hydroxy-2-butanone | 5077-67-8 | 88.11 | 100 MG
SDP

Catalog No. 5000445483
Encompass_Preferred

1-Hydroxy-2-butanone is a natural compound isolated from Bamboo Juice. It acts as a key intermediate for ethambutol, which has anti-tuberculosis effects.

  • Natural compound
  • Key intermediate for ethambutol
  • Anti-tuberculosis effects
  • For research use only
  • Molecular weight: 88.11
  • Purity: 97.38%
  • Formula: C4H8O2
  • Appearance: Liquid, colorless to light yellow

Catalog No. 50-004-45483 Supplier Medchemexpress LLC Supplier No. HYW005327100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Hydroxy-2-butanone is a natural compound isolated from Bamboo Juice. It acts as a key intermediate for ethambutol, which has anti-tuberculosis effects.

  • Natural compound
  • Key intermediate for ethambutol
  • Anti-tuberculosis effects
  • For research use only
  • Molecular weight: 88.11
  • Purity: 97.38%
  • Formula: C4H8O2
  • Appearance: Liquid, colorless to light yellow

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.