Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-Hydroxy-2-butanone | 5077-67-8 | 88.11 | 25 G
SDP

Catalog No. 5000448156
Encompass_Preferred

1-Hydroxy-2-butanone is a natural compound isolated from Bomboo Juice. It serves as a key intermediate for ethambutol, which is known for its anti-tuberculosis effects.

  • Natural compound
  • Key intermediate for ethambutol
  • Exhibits anti-tuberculosis effects
  • Molecular formula: C4H8O2
  • Appearance: Liquid
  • Color: Colorless to light yellow
  • Density: 1.0272 g/cm³
  • Shipping at room temperature (continental US)
  • Recommended storage for pure form: -20°C for 3 years, 4°C for 2 years
  • Recommended storage in solvent: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-004-48156 Supplier Medchemexpress LLC Supplier No. 25GHYW005327
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Hydroxy-2-butanone is a natural compound isolated from Bomboo Juice. It serves as a key intermediate for ethambutol, which is known for its anti-tuberculosis effects.

  • Natural compound
  • Key intermediate for ethambutol
  • Exhibits anti-tuberculosis effects
  • Molecular formula: C4H8O2
  • Appearance: Liquid
  • Color: Colorless to light yellow
  • Density: 1.0272 g/cm³
  • Shipping at room temperature (continental US)
  • Recommended storage for pure form: -20°C for 3 years, 4°C for 2 years
  • Recommended storage in solvent: -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.