Learn More
Medchemexpress LLC 1-Hydroxy-2-butanone | 5077-67-8 | 88.11 | 500 MG
SDP
Catalog No. 5000446047
Encompass_Preferred
1-Hydroxy-2-butanone is a natural compound isolated from Bomboo Juice. It serves as a key intermediate for ethambutol, which is known for its anti-tuberculosis effects.
  • Natural compound
  • Isolated from Bomboo Juice
  • Acts as a key intermediate for ethambutol
  • Ethambutol (derived from this intermediate) has anti-tuberculosis effects
  • Classified as a ketone, aldehyde, and acid
  • Initial source: Microorganisms
  • Endogenous metabolite
Catalog No. 50-004-46047 Supplier Medchemexpress LLC Supplier No. HYW005327500MG
1-Hydroxy-2-butanone is a natural compound isolated from Bomboo Juice. It serves as a key intermediate for ethambutol, which is known for its anti-tuberculosis effects.
  • Natural compound
  • Isolated from Bomboo Juice
  • Acts as a key intermediate for ethambutol
  • Ethambutol (derived from this intermediate) has anti-tuberculosis effects
  • Classified as a ketone, aldehyde, and acid
  • Initial source: Microorganisms
  • Endogenous metabolite